siRNA Details
| Gene Symbol: Ndufv2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031064.2 | NP_112326.1 | 4, 5 | 359 | ||
| GenBank mRNA | BC058495.1 | - | 4, 5 | 363 | 6891104 | |
| M22756.1 | - | 4, 5 | 284 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ndufv2| Gene Symbol: Ndufv2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031064.2 | NP_112326.1 | 4, 5 | 359 | ||
| GenBank mRNA | BC058495.1 | - | 4, 5 | 363 | 6891104 | |
| M22756.1 | - | 4, 5 | 284 | |||