siRNA Details
| Gene Symbol: Rnf123 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032543.2 | NP_115932.1 | 6 | 565 | ||
| GenBank mRNA | AK137599.1 | - | 6 | 510 | ||
| AK157870.1 | - | 6 | 475 | |||
| AK158320.1 | - | 6 | 517 | |||
| AK172244.1 | - | 6 | 489 | |||
| AK177373.1 | - | 2 | 95 | |||
| AY744153.1 | - | 5 | 364 | |||
| BC025530.1 | - | 6 | 486 | |||
| BC057082.1 | - | 6 | 960 | 5704926 | ||

Rnf123
Genomic Map