siRNA Details
| Gene Symbol: D17H6S53E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033477.2 | NP_258438.1 | 3 | 1089 | ||
| GenBank mRNA | BC138707.1 | - | 1 | 785 | 8861729 | |
| BC138708.1 | - | 1 | 785 | 8861730 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

D17H6S53E| Gene Symbol: D17H6S53E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033477.2 | NP_258438.1 | 3 | 1089 | ||
| GenBank mRNA | BC138707.1 | - | 1 | 785 | 8861729 | |
| BC138708.1 | - | 1 | 785 | 8861730 | ||