siRNA Details
| Gene Symbol: OLFM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001288821.1 | NP_001275750.1 | 6 | 2440 | ||
| NM_001288823.1 | NP_001275752.1 | 7 | 2752 | |||
| NM_058170.3 | NP_477518.2 | 6 | 2691 | |||
| NR_110210.1 | 8 | 3489 | ||||
| NR_110211.1 | 7 | 3428 | ||||
| NR_110212.1 | 7 | 3177 | ||||
| XM_017000240.1 | XP_016855729.1 | 7 | 2595 | |||
| GenBank mRNA | AK095551.1 | - | 4 | 1916 | ||
| BC022531.1 | - | 6 | 2450 | 4812035 | ||

OLFM3
Genomic Map