siRNA Details
| Gene Symbol: Pacs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153129.2 | NP_694769.2 | 3 | 688 | ||
| GenBank mRNA | AK154563.1 | - | 561 | |||
| AK164770.1 | - | 4 | 674 | |||
| AK171969.1 | - | 3 | 671 | |||
| BC062886.1 | - | 3 | 688 | 5706409 | ||

Pacs1
Genomic Map