siRNA Details
| Gene Symbol: MT1DP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF348999.2 | - | 4 | 249 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MT1DP| Gene Symbol: MT1DP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF348999.2 | - | 4 | 249 | ||