siRNA Details
Gene Symbol: Krt5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183333.1 | NP_899162.1 | 1939 | |||
GenBank mRNA | AY342389.1 | - | 1939 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Krt5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183333.1 | NP_899162.1 | 1939 | |||
GenBank mRNA | AY342389.1 | - | 1939 |