siRNA Details
| Gene Symbol: Pcdha5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199505.4 | NP_955799.1 | 1 | 740 | ||
| GenBank mRNA | AB113388.1 | - | 1 | 558 | ||
| AY573979.1 | - | 1 | 740 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcdha5| Gene Symbol: Pcdha5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199505.4 | NP_955799.1 | 1 | 740 | ||
| GenBank mRNA | AB113388.1 | - | 1 | 558 | ||
| AY573979.1 | - | 1 | 740 | |||