siRNA Details
Gene Symbol: Olr1666 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000108.1 | NP_001000108.1 | 1 | 474 | ||
XM_006255359.2 | XP_006255421.1 | 2 | 953 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olr1666 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000108.1 | NP_001000108.1 | 1 | 474 | ||
XM_006255359.2 | XP_006255421.1 | 2 | 953 |