siRNA Details
| Gene Symbol: Olr1666 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000108.1 | NP_001000108.1 | 1 | 463 | ||
| XM_006255359.2 | XP_006255421.1 | 2 | 942 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr1666| Gene Symbol: Olr1666 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000108.1 | NP_001000108.1 | 1 | 463 | ||
| XM_006255359.2 | XP_006255421.1 | 2 | 942 | |||