siRNA Details
| Gene Symbol: Olr35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000121.1 | NP_001000121.1 | 1 | 498 | ||
| GenBank mRNA | BC086526.1 | - | 512 | 7303669 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr35| Gene Symbol: Olr35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000121.1 | NP_001000121.1 | 1 | 498 | ||
| GenBank mRNA | BC086526.1 | - | 512 | 7303669 | ||