siRNA Details
| Gene Symbol: Olr122 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000156.1 | NP_001000156.1 | 1 | 851 | ||
| Gene Symbol: LOC100361642 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006229925.1 | XP_006229987.1 | 1 | 851 | ||

Olr122
Genomic Map