siRNA Details
| Gene Symbol: KIAA0196 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014846.3 | NP_055661.3 | 2, 3 | 523 | ||
| XM_011517409.1 | XP_011515711.1 | 2, 3 | 500 | |||
| XM_017014113.1 | XP_016869602.1 | 2, 3 | 728 | |||
| GenBank mRNA | AK223628.1 | - | 2, 3 | 471 | ||
| AK291032.1 | - | 2, 3 | 346 | |||
| BC026951.1 | - | 1, 2 | 274 | 3882443 | ||
| BC106015.1 | - | 2, 3 | 412 | 6059577 | ||
| D83780.1 | - | 2, 3 | 467 | |||

KIAA0196
Genomic Map