siRNA Details
| Gene Symbol: Olr281 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000225.1 | NP_001000225.1 | 1 | 243 | ||
| Gene Symbol: LOC100912217 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003748966.3 | XP_003749014.2 | 1 | 256 | ||
| Gene Symbol: LOC100912408 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003748968.3 | XP_003749016.2 | 1 | 256 | ||

Olr281
Genomic Map