siRNA Details
| Gene Symbol: IQSEC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134382.2 | NP_001127854.1 | 4 | 1692 | ||
| NM_014869.6 | NP_055684.3 | 4 | 1878 | |||
| XM_011534304.2 | XP_011532606.1 | 6 | 1929 | |||
| XM_011534305.2 | XP_011532607.1 | 4 | 1678 | |||
| XM_011534306.2 | XP_011532608.1 | 4 | 2084 | |||
| XM_011534307.2 | XP_011532609.1 | 5 | 2030 | |||
| XM_011534308.2 | XP_011532610.1 | 4 | 1873 | |||
| XM_011534310.2 | XP_011532612.1 | 6 | 1929 | |||
| XM_011534311.2 | XP_011532613.1 | 4 | 2041 | |||
| XM_011534312.2 | XP_011532614.1 | 4 | 1669 | |||
| XM_011534313.2 | XP_011532615.1 | 4 | 1888 | |||
| XM_011534314.2 | XP_011532616.1 | 4 | 1624 | |||
| XM_011534315.2 | XP_011532617.1 | 4 | 1627 | |||
| XM_017007581.1 | XP_016863070.1 | 6 | 1929 | |||
| XM_017007582.1 | XP_016863071.1 | 5 | 1884 | |||
| GenBank mRNA | AB018306.1 | - | 3 | 1371 | ||
| AK294525.1 | - | 4 | 1692 | |||
| AY653734.1 | - | 4 | 1640 | |||
| BC010267.1 | - | 4 | 1632 | 2900367 | ||

IQSEC1
Genomic Map