siRNA Details
| Gene Symbol: Olr607 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000334.1 | NP_001000334.1 | 1 | 931 | ||
| Gene Symbol: LOC100912605 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749515.2 | XP_003749563.1 | 1 | 931 | ||

Olr607
Genomic Map