siRNA Details
| Gene Symbol: Olr1223 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000441.1 | NP_001000441.1 | 1 | 401 | ||
| Gene Symbol: LOC100910822 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008766091.1 | XP_008764313.1 | 401 | |||

Olr1223
Genomic Map