siRNA Details
| Gene Symbol: Olr1096 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000485.1 | NP_001000485.1 | 2 | 815 | ||
| XM_008762647.1 | XP_008760869.1 | 2 | 1354 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr1096| Gene Symbol: Olr1096 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000485.1 | NP_001000485.1 | 2 | 815 | ||
| XM_008762647.1 | XP_008760869.1 | 2 | 1354 | |||