siRNA Details
| Gene Symbol: Olr1687 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000508.1 | NP_001000508.1 | 1 | 789 | ||
| GenBank mRNA | AF091563.1 | - | 1 | 588 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr1687| Gene Symbol: Olr1687 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000508.1 | NP_001000508.1 | 1 | 789 | ||
| GenBank mRNA | AF091563.1 | - | 1 | 588 | ||