siRNA Details
| Gene Symbol: Olr63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000542.1 | NP_001000542.1 | 1 | 797 | ||
| XM_006229890.2 | XP_006229952.1 | 2 | 1114 | |||
| GenBank mRNA | AY834218.1 | - | 1 | 797 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr63| Gene Symbol: Olr63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000542.1 | NP_001000542.1 | 1 | 797 | ||
| XM_006229890.2 | XP_006229952.1 | 2 | 1114 | |||
| GenBank mRNA | AY834218.1 | - | 1 | 797 | ||