siRNA Details
| Gene Symbol: Olr496 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000681.1 | NP_001000681.1 | 1 | 921 | ||
| Gene Symbol: LOC100912505 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749500.1 | XP_003749548.1 | 1 | 921 | ||

Olr496
Genomic Map