siRNA Details
| Gene Symbol: FRMPD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014907.2 | NP_055722.2 | 4, 5 | 414 | ||
| XM_011517803.2 | XP_011516105.1 | 5, 6 | 816 | |||
| XM_011517804.2 | XP_011516106.1 | 4, 5 | 735 | |||
| XM_017014480.1 | XP_016869969.1 | 4, 5 | 1095 | |||
| GenBank mRNA | AB023184.1 | - | 4, 5 | 414 | ||
| BC114965.1 | - | 4, 5 | 393 | 40082639 | ||
| BX648967.1 | - | 4, 5 | 964 | |||

FRMPD1
Genomic Map