siRNA Details
| Gene Symbol: Olr241 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000734.1 | NP_001000734.1 | 1 | 303 | ||
| Gene Symbol: Olr240 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006229963.2 | XP_006230025.1 | 1 | 196 | ||

Olr241
Genomic Map