siRNA Details
| Gene Symbol: Olr86 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000747.1 | NP_001000747.1 | 1 | 224 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr86| Gene Symbol: Olr86 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000747.1 | NP_001000747.1 | 1 | 224 | ||