siRNA Details
| Gene Symbol: Olr811 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000848.1 | NP_001000848.1 | 248 | |||
| Gene Symbol: LOC103692138 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008762957.1 | XP_008761179.1 | 1 | 248 | ||

Olr811
Genomic Map