siRNA Details
| Gene Symbol: Olr804 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000852.1 | NP_001000852.1 | 1 | 22 | ||
| XM_008762900.1 | XP_008761122.1 | 2 | 136 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr804| Gene Symbol: Olr804 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000852.1 | NP_001000852.1 | 1 | 22 | ||
| XM_008762900.1 | XP_008761122.1 | 2 | 136 | |||