siRNA Details
| Gene Symbol: Olr830 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000900.1 | NP_001000900.1 | 1 | 38 | ||
| XM_006237195.1 | XP_006237257.1 | 2 | 109 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr830| Gene Symbol: Olr830 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000900.1 | NP_001000900.1 | 1 | 38 | ||
| XM_006237195.1 | XP_006237257.1 | 2 | 109 | |||