siRNA Details
Gene Symbol: Olr830 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000900.1 | NP_001000900.1 | 1 | 31 | ||
XM_006237195.1 | XP_006237257.1 | 2 | 102 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olr830 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000900.1 | NP_001000900.1 | 1 | 31 | ||
XM_006237195.1 | XP_006237257.1 | 2 | 102 |