siRNA Details
| Gene Symbol: Olr490 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000933.1 | NP_001000933.1 | 1 | 80 | ||
| Gene Symbol: LOC100909796 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749505.1 | XP_003749553.1 | 1 | 80 | ||

Olr490
Genomic Map