siRNA Details
| Gene Symbol: Olr810 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000975.1 | NP_001000975.1 | 29 | |||
| Gene Symbol: LOC103690291 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008762956.1 | XP_008761178.1 | 1 | 38 | ||

Olr810
Genomic Map