siRNA Details
| Gene Symbol: Olr1064 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001076.1 | NP_001001076.1 | 1 | 513 | ||
| Gene Symbol: LOC100910928 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006249611.1 | XP_006249673.1 | 1 | 513 | ||

Olr1064
Genomic Map