siRNA Details
| Gene Symbol: Olr1137 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001079.1 | NP_001001079.1 | 1 | 267 | ||
| Gene Symbol: LOC100911893 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750438.1 | XP_003750486.1 | 1 | 267 | ||

Olr1137
Genomic Map