siRNA Details
| Gene Symbol: AREL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039479.1 | NP_001034568.1 | 9, 10 | 1663 | ||
| XM_005268244.3 | XP_005268301.1 | 8, 9 | 1450 | |||
| XM_006720344.2 | XP_006720407.1 | 9, 10 | 1644 | |||
| XM_011537415.2 | XP_011535717.1 | 10, 11 | 1736 | |||
| XM_011537416.2 | XP_011535718.1 | 9, 10 | 1674 | |||
| XM_011537417.2 | XP_011535719.1 | 9, 10 | 1518 | |||
| GenBank mRNA | AB002315.1 | - | 9, 10 | 1629 | ||
| AK296912.1 | - | 8, 9 | 1421 | |||
| AK304217.1 | - | 9, 10 | 1640 | |||
| BC032944.2 | - | 9, 10 | 1667 | 5267463 | ||

AREL1
Genomic Map