siRNA Details
| Gene Symbol: Lmo7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001515.1 | NP_001001515.1 | 9, 10 | 1196 | ||
| XM_006252397.2 | XP_006252459.1 | 9, 10 | 1432 | |||
| GenBank mRNA | AY533473.1 | - | 9, 10 | 1196 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lmo7| Gene Symbol: Lmo7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001515.1 | NP_001001515.1 | 9, 10 | 1196 | ||
| XM_006252397.2 | XP_006252459.1 | 9, 10 | 1432 | |||
| GenBank mRNA | AY533473.1 | - | 9, 10 | 1196 | ||