siRNA Details
| Gene Symbol: Sucnr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001518.1 | NP_001001518.1 | 1 | 806 | ||
| GenBank mRNA | AY612851.1 | - | 1 | 806 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sucnr1| Gene Symbol: Sucnr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001518.1 | NP_001001518.1 | 1 | 806 | ||
| GenBank mRNA | AY612851.1 | - | 1 | 806 | ||
Set Membership: |
|