siRNA Details
| Gene Symbol: FAM150B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002919.2 | NP_001002919.2 | 2, 3 | 607 | ||
| XM_005264689.4 | XP_005264746.1 | 2, 3 | 620 | |||
| XM_006711879.3 | XP_006711942.1 | 2, 3 | 621 | |||
| XM_011510342.2 | XP_011508644.1 | 2, 3 | 632 | |||
| GenBank mRNA | AY358517.1 | - | 2, 3 | 76 | ||
| BC033490.1 | - | 2, 3 | 481 | |||

FAM150B
Genomic Map