siRNA Details
| Gene Symbol: STUM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003665.3 | NP_001003665.1 | 4 | 1992 | ||
| XM_011544183.2 | XP_011542485.1 | 3 | 2545 | |||
| GenBank mRNA | AL137507.1 | - | 4 | 1992 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

STUM| Gene Symbol: STUM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003665.3 | NP_001003665.1 | 4 | 1992 | ||
| XM_011544183.2 | XP_011542485.1 | 3 | 2545 | |||
| GenBank mRNA | AL137507.1 | - | 4 | 1992 | ||