siRNA Details
| Gene Symbol: TXNDC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003936.3 | NP_001003936.1 | 5 | 356 | ||
| NM_001286946.1 | NP_001273875.1 | 4 | 296 | |||
| XM_017014579.1 | XP_016870068.1 | 5 | 359 | |||
| XM_017014581.1 | XP_016870070.1 | 5 | 359 | |||
| XM_017014582.1 | XP_016870071.1 | 4 | 299 | |||
| XM_017014584.1 | XP_016870073.1 | 5 | 475 | |||
| GenBank mRNA | AF305830.1 | - | 5 | 296 | ||
| BC130549.1 | - | 5 | 286 | 40146580 | ||
| BC130551.1 | - | 5 | 286 | 40146582 | ||
| BC144461.1 | - | 4 | 226 | 9052987 | ||

TXNDC8
Genomic Map