siRNA Details
| Gene Symbol: PALD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014431.2 | NP_055246.2 | 20 | 4330 | ||
| XM_011539637.2 | XP_011537939.1 | 20 | 5516 | |||
| XM_011539638.2 | XP_011537940.1 | 20 | 4103 | |||
| XM_017016072.1 | XP_016871561.1 | 20 | 4101 | |||
| GenBank mRNA | AB033100.2 | - | 20 | 4328 | ||
| AK021806.1 | - | 8 | 1577 | |||
| BC015004.1 | - | 6 | 2191 | |||
| BC040163.1 | - | 18 | 3835 | |||
| BC048195.1 | - | 6 | 2175 | |||

PALD1
Genomic Map