siRNA Details
| Gene Symbol: Lrp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031322.3 | NP_112612.2 | 12 | 1403 | ||
| XM_008762038.1 | XP_008760260.1 | 12 | 1615 | |||
| GenBank mRNA | AB073317.3 | - | 12 | 1403 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrp4| Gene Symbol: Lrp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031322.3 | NP_112612.2 | 12 | 1403 | ||
| XM_008762038.1 | XP_008760260.1 | 12 | 1615 | |||
| GenBank mRNA | AB073317.3 | - | 12 | 1403 | ||