siRNA Details
Gene Symbol: Lrp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031322.3 | NP_112612.2 | 15 | 2061 | ||
XM_008762038.1 | XP_008760260.1 | 15 | 2273 | |||
GenBank mRNA | AB011533.1 | - | 2 | 227 | ||
AB073317.3 | - | 15 | 2061 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
