siRNA Details
| Gene Symbol: Btnl7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212488.1 | NP_997653.1 | 1601 | |||
| XM_008772737.1 | XP_008770959.1 | 1601 | ||||
| Gene Symbol: LOC100911983 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008772783.1 | XP_008771005.1 | 10 | 1790 | ||

Btnl7
Genomic Map