siRNA Details
| Gene Symbol: Wdr46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212491.2 | NP_997656.1 | 15 | 1995 | ||
| GenBank mRNA | BC085934.1 | - | 15 | 1995 | 7112671 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wdr46| Gene Symbol: Wdr46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212491.2 | NP_997656.1 | 15 | 1995 | ||
| GenBank mRNA | BC085934.1 | - | 15 | 1995 | 7112671 | |