siRNA Details
| Gene Symbol: Ier3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212505.2 | NP_997670.1 | 2 | 937 | ||
| GenBank mRNA | BC099831.1 | - | 2 | 913 | 7933804 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ier3| Gene Symbol: Ier3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212505.2 | NP_997670.1 | 2 | 937 | ||
| GenBank mRNA | BC099831.1 | - | 2 | 913 | 7933804 | |