siRNA Details
Gene Symbol: Olr1466 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000024.1 | NP_001000024.1 | 1 | 906 | ||
XM_008767806.1 | XP_008766028.1 | 2 | 2224 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olr1466 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000024.1 | NP_001000024.1 | 1 | 906 | ||
XM_008767806.1 | XP_008766028.1 | 2 | 2224 |