siRNA Details
Gene Symbol: Olr1475 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000027.1 | NP_001000027.1 | 1 | 902 |
Gene Symbol: LOC100909438 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003752201.2 | XP_003752249.1 | 1 | 906 |
