siRNA Details
| Gene Symbol: PPP1R1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006741.3 | NP_006732.3 | 7 | 1071 | ||
| GenBank mRNA | BC022470.1 | - | 7 | 1071 | 4819907 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

PPP1R1A| Gene Symbol: PPP1R1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006741.3 | NP_006732.3 | 7 | 1071 | ||
| GenBank mRNA | BC022470.1 | - | 7 | 1071 | 4819907 | |