siRNA Details
Gene Symbol: GPR75 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006794.3 | NP_006785.1 | 2 | 541 | ||
GenBank mRNA | AF072693.1 | - | 2 | 436 | ||
AK314885.1 | - | 2 | 497 | |||
BC067475.1 | - | 1 | 337 | 6971705 | ||
BC067476.1 | - | 1 | 258 | 6971810 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
