siRNA Details
| Gene Symbol: Olr128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000159.1 | NP_001000159.1 | 1 | 916 | ||
| Gene Symbol: LOC103694854 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773798.1 | XP_008772020.1 | 1 | 949 | ||

Olr128
Genomic Map