siRNA Details
| Gene Symbol: Olr598 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000332.1 | NP_001000332.1 | 271 | |||
| Gene Symbol: LOC100911380 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006234490.2 | XP_006234552.1 | 1 | 271 | ||

Olr598
Genomic Map